4-Bromo-5-fluoro-2-nitrobenzoic acid ≥95%
Supplier: Matrix Scientific
072688-1G
072688-5G
102526-192EA
52.31
USD
102526-192
102526-194
4-Bromo-5-fluoro-2-nitrobenzoic acid ≥95%
4-Bromo-5-fluoro-2-nitrobenzoic acid
Hazard: Irritant
Formula:
C₇H₃BrFNO₄ MW: 264.01 g/mol |
MDL Number:
MFCD08702773 CAS Number: 1020717-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...