2-Amino-3-methoxypyridine ≥95%

Supplier: Matrix Scientific

072687-1G 072687-5G
102526-188EA 78.46 USD
102526-188 102526-190
2-Amino-3-methoxypyridine ≥95%
2-Amino-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
MDL Number: MFCD07374874
CAS Number: 10201-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR