2-Hydroxy-5-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methyl-2-pyridone, 2-hydroxy-5-picoline

072684-5G 072684-100G 072684-25G
102526-172EA 118.46 USD
102526-172 102526-176 102833-536
2-Hydroxy-5-methylpyridine ≥95%
2-Hydroxy-5-methylpyridine

Hazard: Irritant

Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD03092888
CAS Number: 1003-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR