4-(4-Hydroxyphenyl)butanoic acid ≥95%

Supplier: Matrix Scientific

072680-1G 072680-5G
102526-152EA 238.46 USD
102526-152 102526-154
4-(4-Hydroxyphenyl)butanoic acid ≥95%
4-(4-Hydroxyphenyl)butanoic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₃
Melting Pt: 110…111 °C
MDL Number: MFCD00033287
CAS Number: 7021-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR