2-(5-Bromopyrimidin-2-yl)acetonitrile ≥95%

Supplier: Matrix Scientific

072676-250MG 072676-1G
102526-132EA 739.51 USD
102526-132 102526-134
2-(5-Bromopyrimidin-2-yl)acetonitrile ≥95%
2-(5-Bromopyrimidin-2-yl)acetonitrile

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
MDL Number: MFCD11111850
CAS Number: 831203-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR