1-tert-Butyl-6-methyl-1H-indole-1,6-dicarboxylate ≥95%

Supplier: Matrix Scientific

072675-250MG 072675-1G 072675-5G
102526-126EA 250.11 USD
102526-126 102526-128 102526-130
1-tert-Butyl-6-methyl-1H-indole-1,6-dicarboxylate ≥95%
1-tert-Butyl-6-methyl-1H-indole-1,6-dicarboxylate

Hazard: Irritant

Formula: C₁₅H₁₇NO₄
CAS Number: 957127-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR