2-Fluoronicotinamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoropyridine-3-carboxamide

072674-250MG 072674-1G 072674-5G
102526-120EA 169.23 USD
102526-120 102526-122 102526-124
2-Fluoronicotinamide ≥95%
2-Fluoronicotinamide

Hazard: Irritant

Formula: C₆H₅FN₂O
MW: 140.115 g/mol
MDL Number: MFCD11100137
CAS Number: 364-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR