Thiazole-5-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Cyanothiazole

072673-250MG 072673-1G
102526-116EA 967.69 USD
102526-116 102526-118
Thiazole-5-carbonitrile ≥95%
Thiazole-5-carbonitrile

Hazard: Irritant

Formula: C₄H₂N₂S
MW: 110.1371 g/mol
MDL Number: MFCD06797206
CAS Number: 25742-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR