2,2-Dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one ≥95%

Supplier: Matrix Scientific

072671-250MG 072671-1G 072671-5G
102526-106EA 640 USD
102526-106 102526-108 102526-110
2,2-Dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one ≥95%
2,2-Dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MW: 178.1879 g/mol
MDL Number: MFCD10000810
CAS Number: 20348-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR