6-Bromo-1,2,3,4-tetrahydroisoquinoline ≥95%

Supplier: Matrix Scientific

072670-1G 072670-5G
102526-102EA 75.38 USD
102526-102 102526-104
6-Bromo-1,2,3,4-tetrahydroisoquinoline ≥95%
6-Bromo-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₉H₁₀BrN
MDL Number: MFCD07374370
CAS Number: 226942-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR