N4-(3-Bromophenyl)quinazoline-4,6-diamine ≥95%

Supplier: Matrix Scientific

072668-250MG 072668-1G
102526-094EA 1395.38 USD
102526-094 102526-096
N4-(3-Bromophenyl)quinazoline-4,6-diamine ≥95%
N4-(3-Bromophenyl)quinazoline-4,6-diamine

Hazard: Irritant

Formula: C₁₄H₁₁BrN₄
MDL Number: MFCD09908082
CAS Number: 169205-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR