Methyl-2-fluoronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-fluoronicotinate, 2-Fluoronicotinic acid methyl ester

072665-5G 072665-25G
102526-078EA 98.46 USD
102526-078 102551-800
Methyl-2-fluoronicotinate ≥95%
Methyl-2-fluoronicotinate

Hazard: Irritant/Refrigerate

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 75°C/5mmHg
Melting Pt: 22-26°C
Flash Pt: 110°C
MDL Number: MFCD03095072
CAS Number: 446-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR