4-Chloro-5-methylpyrimidine ≥95%

Supplier: Matrix Scientific

072663-250MG 072663-1G 072663-5G
102526-064EA 720 USD
102526-064 102526-066 102526-068
4-Chloro-5-methylpyrimidine ≥95%
4-Chloro-5-methylpyrimidine

Hazard: Irritant

Formula: C₅H₅ClN₂
MDL Number: MFCD08236809
CAS Number: 51957-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR