2-Chloro-5-fluoropyrimidin-4-one ≥95%

Supplier: Matrix Scientific

072662-1G 072662-5G
102526-060EA 66.15 USD
102526-060 102526-062
2-Chloro-5-fluoropyrimidin-4-one ≥95%
2-Chloro-5-fluoropyrimidin-4-one

Hazard: Irritant

Formula: C₄H₂ClFN₂O
Melting Pt: 176…177 °C
MDL Number: MFCD01346689
CAS Number: 155-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR