5-Methoxypyrazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

072661-250MG 072661-1G 072661-5G
102526-054EA 126.15 USD
102526-054 102526-056 102526-058
5-Methoxypyrazine-2-carboxylic acid ≥95%
5-Methoxypyrazine-2-carboxylic acid

Hazard: Irritant

Formula: C6H6N2O3
MDL Number: MFCD10697773
CAS Number: 40155-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR