2-Fluoro-3-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

072656-250MG 072656-1G
102526-028EA 783.08 USD
102526-028 102526-030
2-Fluoro-3-nitrobenzaldehyde ≥95%
2-Fluoro-3-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₄FNO₃
MDL Number: MFCD08669885
CAS Number: 96516-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR