6-Methylpyridine-3-sulfonic acid ≥95%

Supplier: Matrix Scientific

072655-250MG 072655-1G 072655-5G
102526-022EA 498.46 USD
102526-022 102526-024 102526-026
6-Methylpyridine-3-sulfonic acid ≥95%
6-Methylpyridine-3-sulfonic acid

Hazard: Irritant

Formula: C₆H₇NO₃S
MW: 173.1897 g/mol
MDL Number: MFCD01646153
CAS Number: 4808-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR