2-Chloro-3-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

072654-250MG 072654-1G
102526-018EA 209.23 USD
102526-018 102526-020
2-Chloro-3-nitrobenzaldehyde ≥95%
2-Chloro-3-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₄ClNO₃
MDL Number: MFCD08236804
CAS Number: 58755-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR