tert-Butyl-3,5-dimethylpiperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-3,5-dimethylpiperazine

072651-1G 072651-5G
102526-004EA 183.08 USD
102526-004 102526-006
tert-Butyl-3,5-dimethylpiperazine-1-carboxylate ≥95%
tert-Butyl-3,5-dimethylpiperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD07371499
CAS Number: 639068-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR