2-Chloro-3-fluoro-5-hydroxypyridine ≥95%
Supplier: Matrix Scientific
072650-250MG
072650-1G
072650-5G
102525-998EA
400
USD
102525-998
102526-000
102526-002
2-Chloro-3-fluoro-5-hydroxypyridine ≥95%
2-Chloro-3-fluoro-5-hydroxypyridine
Hazard: Irritant
Formula:
C₆H₄ClFO |
MDL Number:
MFCD03411573 CAS Number: 870062-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...