4-Boc-3-morpholinecarboxylic acid ≥95%

Supplier: Matrix Scientific

072642-1G 072642-5G
102525-962EA 533.85 USD
102525-962 102525-964
4-Boc-3-morpholinecarboxylic acid ≥95%
4-Boc-3-Morpholinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MW: 231.246 g/mol
MDL Number: MFCD02683057
CAS Number: 212650-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR