3,5-Dibromo-6-chloropyrazin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-3,5-dibromo-6-chloropyrazine

072641-1G 072641-5G 072641-10G
102525-956EA 533.85 USD
102525-956 102525-958 102525-960
3,5-Dibromo-6-chloropyrazin-2-amine ≥95%
3,5-Dibromo-6-chloropyrazin-2-amine

Hazard: Irritant

Formula: C4H2Br2ClN3
MW: 287.3400 g/mol
MDL Number: MFCD11043607
CAS Number: 566205-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR