6-Bromo-2-phenyl-1H-imidazo[4,5-b]pyridine ≥95%

Supplier: Matrix Scientific

072639-250MG 072639-1G 072639-5G
102525-944EA 498.46 USD
102525-944 102525-946 102525-948
6-Bromo-2-phenyl-1H-imidazo[4,5-b]pyridine ≥95%
6-Bromo-2-phenyl-1H-imidazo[4,5-b]pyridine

Hazard: Irritant

Formula: C₁₂H₈BrN₃
MW: 274.12 g/mol
MDL Number: MFCD03081184
CAS Number: 65147-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR