7-(Benzyloxy)-4-chloro-6-methoxyquinazoline ≥95%

Supplier: Matrix Scientific

072638-250MG 072638-1G
102525-940EA 476.92 USD
102525-940 102525-942
7-(Benzyloxy)-4-chloro-6-methoxyquinazoline ≥95%
7-(Benzyloxy)-4-chloro-6-methoxyquinazoline

Hazard: Irritant

Formula: C₁₆H₁₃ClN₂O₂
MW: 300.7396 g/mol
MDL Number: MFCD04115120
CAS Number: 162364-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR