5-Bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-phenyl-7-azaindole

072637-250MG 072637-1G
102525-936EA 924.62 USD
102525-936 102525-938
5-Bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine ≥95%
5-Bromo-2-phenyl-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₃H₉BrN₂
MW: 273.12
MDL Number: MFCD11109822
CAS Number: 953414-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR