Methyl-5-bromopyrimidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromopyrimidine-4-carboxylate

072635-250MG 072635-1G 072635-5G
102525-924EA 483.85 USD
102525-924 102525-926 102525-928
Methyl-5-bromopyrimidine-4-carboxylate ≥95%
Methyl-5-bromopyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.0200 g/mol
MDL Number: MFCD11111841
CAS Number: 1009826-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR