2-Bromo-5-cyano-3-picoline ≥95%

Supplier: Matrix Scientific

072633-250MG 072633-1G
102525-912EA 338.46 USD
102525-912 102525-914
2-Bromo-5-cyano-3-picoline ≥95%
2-Bromo-5-cyano-3-picoline

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
MDL Number: MFCD03095063
CAS Number: 374633-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR