3-Cyano-4-oxopiperidine-1-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-cyano-4-oxopiperidine-1-carboxylate

072632-1G 072632-5G
102525-908EA 78.46 USD
102525-908 102525-910
3-Cyano-4-oxopiperidine-1-carboxylic acid tert-butyl ester ≥95%
3-Cyano-4-oxopiperidine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃
MW: 224.26 g/mol
MDL Number: MFCD11109814
CAS Number: 914988-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR