7-Chloro-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Chloro-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine

072630-250MG 072630-1G 072630-5G
102525-894EA 427.69 USD
102525-894 102525-896 102525-898
7-Chloro-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine ≥95%
7-Chloro-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C₆H₅ClN₄
MW: 168.5837 g/mol
MDL Number: MFCD00227619
CAS Number: 24415-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR