tert-Butyl-1-oxa-5-azaspiro[2,5]octane-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 1-oxa-5-azaspiro[2,5]octane-5-carboxylate

072629-250MG 072629-1G 072629-5G
102525-888EA 456.92 USD
102525-888 102525-890 102525-892
tert-Butyl-1-oxa-5-azaspiro[2,5]octane-5-carboxylate ≥95%
tert-Butyl-1-oxa-5-azaspiro[2,5]octane-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MW: 213.2735 g/mol
MDL Number: MFCD07779386
CAS Number: 276872-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR