2-Chlorothieno[3,2-d]pyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

072628-250MG 072628-1G 072628-5G
102525-882EA 640 USD
102525-882 102525-884 102525-886
2-Chlorothieno[3,2-d]pyrimidin-4-amine ≥95%
2-Chlorothieno[3,2-d]pyrimidin-4-amine

Hazard: Irritant

Formula: C₆H₄ClN₃S
MDL Number: MFCD11109812
CAS Number: 16234-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR