5-Methylpyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

072626-250MG 072626-1G 072626-5G
102525-870EA 400 USD
102525-870 102525-872 102525-874
5-Methylpyrimidin-2-amine ≥95%
5-Methylpyrimidin-2-amine

Hazard: Irritant

Formula: C5H7N3
MW: 109.1300 g/mol
MDL Number: MFCD00463477
CAS Number: 50840-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR