Methyl-2-chloropyrimidine-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloropyrimidine-5-carboxylate

072625-1G 072625-5G
102525-866EA 152.31 USD
102525-866 102525-868
Methyl-2-chloropyrimidine-5-carboxylate ≥95%
Methyl-2-chloropyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₆H₅ClN₂O₂
MW: 172.5691 g/mol
MDL Number: MFCD08690330
CAS Number: 287714-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR