2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one ≥95%

Supplier: Matrix Scientific

072621-1G 072621-5G
102525-842EA 427.69 USD
102525-842 102525-844
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one ≥95%
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one

Hazard: Irritant

Formula: C₆H₆N₄O
MW: 150.14 g/mol
MDL Number: MFCD09033259
CAS Number: 7355-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR