4-Bromo-6-(trifluoromethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

072616-1G 072616-5G
102525-816EA 335.38 USD
102525-816 102525-818
4-Bromo-6-(trifluoromethyl)pyrimidine ≥95%
4-Bromo-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₅H₂BrF₃N₂
MW: 226.9800 g/mol
MDL Number: MFCD10574962
CAS Number: 785777-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR