6-Bromo-2-methylbenzo[d]thiazole ≥95%

Supplier: Matrix Scientific

072615-1G 072615-5G
102525-810EA 76.92 USD
102525-810 102525-812
6-Bromo-2-methylbenzo[d]thiazole ≥95%
6-Bromo-2-methylbenzo[d]thiazole

Hazard: Irritant

Formula: C₈H₆BrNS
MDL Number: MFCD00466580
CAS Number: 5304-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR