Pyrimidine-4-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Cyanopyrimidine

072614-250MG 072614-1G 072614-5G
102525-802EA 1295.38 USD
102525-802 102525-804 102525-806
Pyrimidine-4-carbonitrile ≥95%
Pyrimidine-4-carbonitrile

Hazard: Irritant

Formula: C5H3N3
MDL Number: MFCD07787613
CAS Number: 42839-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR