4-Bromo-1-naphthylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Amino-4-bromonaphthalene

072613-5G 072613-10G
102525-798EA 27.69 USD
102525-798 102525-800
4-Bromo-1-naphthylamine ≥95%
4-Bromo-1-naphthylamine

Hazard: Irritant

Formula: C₁₀H₈BrN
MW: 222.08 g/mol
Melting Pt: 100…103 °C
MDL Number: MFCD00004023
CAS Number: 2298-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR