2-Bromo-6-cyanopyrazine ≥95%

Supplier: Matrix Scientific

072612-250MG 072612-1G 072612-5G
102525-792EA 1984.62 USD
102525-792 102525-794 102525-796
2-Bromo-6-cyanopyrazine ≥95%
2-Bromo-6-cyanopyrazine

Hazard: Irritant

Formula: C₅H₂BrN₃
MW: 183.9900 g/mol
MDL Number: MFCD11109800
CAS Number: 859064-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR