2-Amino-6-cyanopyrazine ≥95%
Supplier: Matrix Scientific
072611-1G
072611-5G
102525-788EA
853.85
USD
102525-788
102525-790
2-Amino-6-cyanopyrazine ≥95%
2-Amino-6-cyanopyrazine
Hazard: Irritant
Formula:
C₅H₄N₄ MW: 120.1100 g/mol |
MDL Number:
MFCD10697805 CAS Number: 59489-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...