6-Chloropyrazine-2-carbonitrile ≥95%

Supplier: Matrix Scientific

072610-1G 072610-5G
102525-784EA 130.77 USD
102525-784 102525-786
6-Chloropyrazine-2-carbonitrile ≥95%
6-Chloropyrazine-2-carbonitrile

Hazard: Irritant

Formula: C₅H₂ClN₃
MW: 139.5400 g/mol
MDL Number: MFCD09607699
CAS Number: 6863-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR