7-Bromo-4-chloroquinoline ≥95%

Supplier: Matrix Scientific

072607-1G 072607-5G
102525-770EA 183.08 USD
102525-770 102525-772
7-Bromo-4-chloroquinoline ≥95%
7-Bromo-4-chloroquinoline

Hazard: Irritant

Formula: C₉H₅BrClN
MW: 242.4997 g/mol
MDL Number: MFCD02684221
CAS Number: 75090-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR