8-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥95%

Supplier: Matrix Scientific

072604-1G 072604-5G
102525-754EA 258.46 USD
102525-754 102525-756
8-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥95%
8-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride

Hazard: Irritant

Formula: C₉H₁₁BrClN
MW: 248.5500 g/mol
MDL Number: MFCD08752231
CAS Number: 1159813-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR