6-Bromo-2,3-dihydroquinolin-4(1H)-one ≥95%

Supplier: Matrix Scientific

072602-250MG 072602-1G 072602-5G
102525-742EA 333.85 USD
102525-742 102525-744 102525-746
6-Bromo-2,3-dihydroquinolin-4(1H)-one ≥95%
6-Bromo-2,3-dihydroquinolin-4(1H)-one

Hazard: Irritant

Formula: C₉H₈BrNO
MW: 226.0700 g/mol
MDL Number: MFCD00600620
CAS Number: 76228-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR