2-Amino-4-bromo-6-methylpyrimidine ≥95%
Supplier: Matrix Scientific
072601-250MG
072601-1G
072601-5G
102525-736EA
498.46
USD
102525-736
102525-738
102525-740
2-Amino-4-bromo-6-methylpyrimidine ≥95%
2-Amino-4-bromo-6-methylpyrimidine
Hazard: Irritant
Formula:
C₅H₆BrN₃ |
MDL Number:
MFCD11109799 CAS Number: 5734-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...