4-Amino-6-bromopyrimidine ≥95%

Supplier: Matrix Scientific

072600-1G 072600-5G
102525-732EA 155.38 USD
102525-732 102525-734
4-Amino-6-bromopyrimidine ≥95%
4-Amino-6-bromopyrimidine

Hazard: Irritant

Formula: C₄H₄BrN₃
MDL Number: MFCD00233963
CAS Number: 1159818-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR