4-Amino-2-bromopyrimidine ≥95%

Supplier: Matrix Scientific

072599-250MG 072599-1G 072599-5G
102525-724EA 640 USD
102525-724 102525-726 102525-728
4-Amino-2-bromopyrimidine ≥95%
4-Amino-2-bromopyrimidine

Hazard: Irritant

Formula: C₄H₄BrN₃
MW: 174.0000 g/mol
MDL Number: MFCD11109798
CAS Number: 1160994-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR