3-Fluoro-5-nitroaniline ≥95%

Supplier: Matrix Scientific

072598-1G 072598-5G 072598-10G
102525-718EA 569.23 USD
102525-718 102525-720 102525-722
3-Fluoro-5-nitroaniline ≥95%
3-Fluoro-5-nitroaniline

Hazard: Irritant

Formula: C₆H₅FN₂O₂
MW: 156.12
Melting Pt: 115…116 °C
CAS Number: 2369-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR