4-Amino-3-bromoisoquinoline ≥95%

Supplier: Matrix Scientific

072595-250MG 072595-1G 072595-5G
102525-702EA 640 USD
102525-702 102525-704 102525-706
4-Amino-3-bromoisoquinoline ≥95%
4-Amino-3-bromoisoquinoline

Hazard: Irritant

Formula: C₉H₇BrN₂
MDL Number: MFCD00234504
CAS Number: 40073-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR