Methyl-7-bromo-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 7-bromo-1H-indole-2-carboxylate

072594-250MG 072594-1G 072594-5G
102525-696EA 244.62 USD
102525-696 102525-698 102525-700
Methyl-7-bromo-1H-indole-2-carboxylate ≥95%
Methyl-7-bromo-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈BrNO₂
MW: 254.0800 g/mol
MDL Number: MFCD11111832
CAS Number: 1158503-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR